
trans-Oxyresveratrol

Synonym(s):
Oxyresveratrol, (E)-2,3′,4,5′-Stilbenetetrol, 2,3′,4,5′-Tetrahydroxy-trans-stilbene, 4-[(1E)-2-(3,5-Dihydroxyphenyl)ethenyl]-1,3-benzenediol
Empirical Formula (Hill Notation):
C14H12O4
CAS Number:
Molecular Weight:
244.24
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1