Skip to Content
Merck
Torsemide Related Compound B certified reference material, pharmaceutical secondary standard

Torsemide Related Compound B

Torsemide Related Compound B certified reference material, pharmaceutical secondary standard
Synonym(s):
N-[(Butylamino)carbonyl]-4-[(3-methylphenyl)amino]-3-pyridinesulfonamide
Empirical Formula (Hill Notation):
C17H22N4O3S
CAS Number:
Molecular Weight:
362.45

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade