Tiamulin Related Compound A
Synonym(s):
Tosyl pleuromutilin
Empirical Formula (Hill Notation):
C29H40O7S
CAS Number:
Molecular Weight:
532.69
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2