![Sulfosulfuron PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/902/982/f40ebc30-880c-4971-9ba2-84ebcf3c7578/640/f40ebc30-880c-4971-9ba2-84ebcf3c7578.png)
Sulfosulfuron
![Sulfosulfuron PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/902/982/f40ebc30-880c-4971-9ba2-84ebcf3c7578/640/f40ebc30-880c-4971-9ba2-84ebcf3c7578.png)
Synonym(s):
1-(4,6-Dimethoxypyrimidin-2-yl)-3-(2-ethylsulfonylimidazol[1,2-a]pyridin-3-ylsulfonyl)urea
Empirical Formula (Hill Notation):
C16H18N6O7S2
CAS Number:
Molecular Weight:
470.48
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2