Skip to Content
Merck
vibrant-m

Sotalol Related Compound C

vibrant-m
Synonym(s):
Deoxysotalol hydrochloride
Empirical Formula (Hill Notation):
C12H20N2O2S · HCl
CAS Number:
Molecular Weight:
292.83

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1