Skip to Content
Merck
Sodium periodate ACS reagent, ≥99.8%

Sodium periodate

Sodium periodate ACS reagent, ≥99.8%
Synonym(s):
Sodium (meta)periodate
Linear Formula:
NaIO4
CAS Number:
Molecular Weight:
213.89
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing