![Schizandrin A phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/171/753/e6db2e75-125e-4e71-a217-fcf23a4790a1/640/e6db2e75-125e-4e71-a217-fcf23a4790a1.png)
Schizandrin A
![Schizandrin A phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/171/753/e6db2e75-125e-4e71-a217-fcf23a4790a1/640/e6db2e75-125e-4e71-a217-fcf23a4790a1.png)
Synonym(s):
Schisandrin A, Deoxyschisandrin, Deoxyschizandrin, Schizandrin A
Empirical Formula (Hill Notation):
C24H32O6
CAS Number:
Molecular Weight:
416.51
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing