Skip to Content
Merck
Scandium foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%

Scandium

Scandium foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Synonym(s):
Scandium
Empirical Formula (Hill Notation):
Sc
CAS Number:
Molecular Weight:
44.96

Products

Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Element