Rosarin
Synonym(s):
Rosarin, (2E)-3-Phenyl-2-propen-1-yl 6-O-a-L-arabinofuranosyl-β-D-glucopyranoside
Empirical Formula (Hill Notation):
C20H28O10
CAS Number:
Molecular Weight:
428.43
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing