![(R)-Naproxen pharmaceutical impurity standard](/deepweb/assets/sigmaaldrich/product/structures/152/357/a173f9cb-503a-407e-91a8-6af39c9aeb64/640/a173f9cb-503a-407e-91a8-6af39c9aeb64.png)
(R)-Naproxen
![(R)-Naproxen pharmaceutical impurity standard](/deepweb/assets/sigmaaldrich/product/structures/152/357/a173f9cb-503a-407e-91a8-6af39c9aeb64/640/a173f9cb-503a-407e-91a8-6af39c9aeb64.png)
Synonym(s):
(R)-(−)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, (2R)-2-(6-Methoxy-2-naphthyl)propionic acid, (2R)-2-(6-Methoxynaphthalen-2-yl)propanoic acid, (R)-Naproxen
Linear Formula:
CH3OC10H6CH(CH3)CO2H
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing