Skip to Content
Merck
vibrant-m

Residual Solvent Class 2 - Ethylene Glycol

vibrant-m
Synonym(s):
Ethylene glycol solution
Empirical Formula (Hill Notation):
C2H6O2
CAS Number:
Molecular Weight:
62.07

Products

Formula Weight
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1