Skip to Content
Merck
Prothiofos certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Prothiofos

Prothiofos certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Empirical Formula (Hill Notation):
C11H15Cl2O2PS2
CAS Number:
Molecular Weight:
345.25

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Reference Material Type
Special Grade