Prosulfocarb sulfoxide
Synonym(s):
1-[(Phenylmethyl)sulfinyl]-N,N-dipropylformamide
Empirical Formula (Hill Notation):
C14H21NO2S
CAS Number:
Molecular Weight:
267.39
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing