Skip to Content
Merck
Prenyl formate ≥95%, FG

Prenyl formate

Prenyl formate ≥95%, FG
Synonym(s):
3-methylbut-2-enyl formate
Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14

Products

Product Category
Brand
Formula Weight
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing