Skip to Content
Merck
Paroxetine Related Compound C pharmaceutical secondary standard, certified reference material

Paroxetine Related Compound C

Paroxetine Related Compound C pharmaceutical secondary standard, certified reference material
Synonym(s):
(3R-trans)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine hydrochloride hemihydrate
Empirical Formula (Hill Notation):
C19H20FNO3 · HCl · 0.5H2O
Molecular Weight:
374.83

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade