
Ondansetron impurity E

Synonym(s):
Imidazole, 1,3-Diaza-2,4-cyclopentadiene, Glyoxaline
Empirical Formula (Hill Notation):
C3H4N2
CAS Number:
Molecular Weight:
68.08
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
pK Value
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing