Naphthol AS-BI N-acetyl-β-D-glucosaminide
Synonym(s):
2-Naphthalenecarboxamide, 3-[[2-(acetylamino)-2-deoxy-β-D-glucopyranosyl]oxy]-7-bromo-N-(2-methoxyphenyl)-, Naphthol AS-BI 2-acetamido-2-deoxy-β-D-glucopyranoside
Empirical Formula (Hill Notation):
C26H27BrN2O8
CAS Number:
Molecular Weight:
575.41
Products
Product Category
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing