Monoclonal Anti-HDAC1 antibody produced in mouse
Synonym(s):
GON-10, HD1, RPD3L1, Anti-DKFZp686H12203, Anti-GON10, Anti-HD1, Anti-RPD3, Anti-RPD3L1
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing