![Memantine Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/426/165/8593a2d7-8e91-4cff-8a29-cacd33ddbe9f/640/8593a2d7-8e91-4cff-8a29-cacd33ddbe9f.png)
Memantine Related Compound C
![Memantine Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/426/165/8593a2d7-8e91-4cff-8a29-cacd33ddbe9f/640/8593a2d7-8e91-4cff-8a29-cacd33ddbe9f.png)
Synonym(s):
1-Chloro-3,5-dimethyladamantane
Empirical Formula (Hill Notation):
C12H19Cl
CAS Number:
Molecular Weight:
198.73
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2