Skip to Content
Merck
Lidocaine British Pharmacopoeia (BP) Reference Standard

Lidocaine

Lidocaine British Pharmacopoeia (BP) Reference Standard
Synonym(s):
2-Diethylamino-N-(2,6-dimethylphenyl)acetamide, Lignocaine, Xylocaine
Empirical Formula (Hill Notation):
C14H22N2O
CAS Number:
Molecular Weight:
234.34
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique