![Leptophos certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/179/871/fca3b7c3-22b1-4c65-8c7a-d9d82189f112/640/fca3b7c3-22b1-4c65-8c7a-d9d82189f112.png)
Leptophos
![Leptophos certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/179/871/fca3b7c3-22b1-4c65-8c7a-d9d82189f112/640/fca3b7c3-22b1-4c65-8c7a-d9d82189f112.png)
Synonym(s):
O-(4-Bromo-2,5-dichlorophenyl) O-methyl phenylphosphonothioate
Empirical Formula (Hill Notation):
C13H10BrCl2O2PS
CAS Number:
Molecular Weight:
412.07
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2