Lansoprazole
Synonym(s):
Lansoprazole
Empirical Formula (Hill Notation):
C16H14F3N3O2S
CAS Number:
Molecular Weight:
369.36
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing