Skip to Content
Merck
Lactic acid United States Pharmacopeia (USP) Reference Standard

Lactic acid

Lactic acid United States Pharmacopeia (USP) Reference Standard
Synonym(s):
DL-Lactic acid, 2-Hydroxypropionic acid
Linear Formula:
CH3CH(OH)COOH
CAS Number:
Molecular Weight:
90.08
EC Number:

Products

Shipping
Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique