Skip to Content
Merck
Isophorone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Isophorone

Isophorone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
3,5,5-Trimethyl-2-cyclohexen-1-one
Empirical Formula (Hill Notation):
C9H14O
CAS Number:
Molecular Weight:
138.21

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade