Homoorientin
Synonym(s):
Isoorientin, Homoorientin, Luteolin 6-C-β-D-glucoside, Luteolin 6-C-glucoside
Empirical Formula (Hill Notation):
C21H20O11
CAS Number:
Molecular Weight:
448.38
Products
Shipping
Formula Weight
Manufacturer
Physical Form
Product Line
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing