Skip to Content
Merck
Glyceryl tributyrate analytical standard

Glyceryl tributyrate

Glyceryl tributyrate analytical standard
Synonym(s):
1,2,3-Tributyrylglycerol, Glycerol tributyrate, Tributyrin
Linear Formula:
(CH3CH2CH2COOCH2)2CHOCOCH2CH2CH3
CAS Number:
Molecular Weight:
302.36
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing