Skip to Content
Merck
vibrant-m

Gliclazide impurity F

vibrant-m
Synonym(s):
N-[[(Hexahydrocyclopenta[c]pyrrol-2(1H)-yl)amino]carbonyl]-2-methylbenzenesulfonamide, 1-(Hexahydrocyclopenta[c]pyrrol-2(1H)-yl)-3-[(2-methylphenyl)sulphonyl]urea
Empirical Formula (Hill Notation):
C15H21N3O3S
CAS Number:
Molecular Weight:
323.41

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade