Skip to Content
Merck
Fmoc-Thr(tBu)-OH ≥98.0% (HPLC)

Fmoc-Thr(tBu)-OH

Fmoc-Thr(tBu)-OH ≥98.0% (HPLC)
Synonym(s):
Fmoc-O-tert-butyl-L-threonine, Fmoc-Thr(tBu)-OH
Empirical Formula (Hill Notation):
C23H27NO5
CAS Number:
Molecular Weight:
397.46

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing