Skip to Content
Merck
vibrant-m

Fludarabine for system suitability

vibrant-m
Synonym(s):
Fludarabine phosphate, 2-Fluoro-9-(5-O-phosphono-β-D-arabinofuranosyl)-9H-purin-6-amine
Empirical Formula (Hill Notation):
C10H13FN5O7P
CAS Number:
Molecular Weight:
365.21

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1