Skip to Content
Merck
Fenofibrate European Pharmacopoeia (EP) Reference Standard

Fenofibrate

Fenofibrate European Pharmacopoeia (EP) Reference Standard
Synonym(s):
2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoic acid isopropyl ester
Empirical Formula (Hill Notation):
C20H21ClO4
CAS Number:
Molecular Weight:
360.83

Products

Shipping
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique