Skip to Content
Merck
Exemestane European Pharmacopoeia (EP) Reference Standard

Exemestane

Exemestane European Pharmacopoeia (EP) Reference Standard
Synonym(s):
6-Methyleneandrosta-1,4-diene-3,17-dione, Exemestane
Empirical Formula (Hill Notation):
C20H24O2
CAS Number:
Molecular Weight:
296.40

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique