![Ethylene sulfate ≥99%, anhydrous, battery grade](/deepweb/assets/sigmaaldrich/product/structures/367/849/75985e77-de5c-47f6-99f4-f837e08df1be/640/75985e77-de5c-47f6-99f4-f837e08df1be.png)
Ethylene sulfate
![Ethylene sulfate ≥99%, anhydrous, battery grade](/deepweb/assets/sigmaaldrich/product/structures/367/849/75985e77-de5c-47f6-99f4-f837e08df1be/640/75985e77-de5c-47f6-99f4-f837e08df1be.png)
Synonym(s):
1,3,2-Dioxathiolane 2,2-dioxide
Empirical Formula (Hill Notation):
C2H4O4S
CAS Number:
Molecular Weight:
124.12
Products
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing