![(E)-β-Damascone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/240/444/a6f4c41a-ae09-4940-8d79-2aefcf44c50d/640/a6f4c41a-ae09-4940-8d79-2aefcf44c50d.png)
(E)-β-Damascone
![(E)-β-Damascone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/240/444/a6f4c41a-ae09-4940-8d79-2aefcf44c50d/640/a6f4c41a-ae09-4940-8d79-2aefcf44c50d.png)
Synonym(s):
(E)-1-(2,6,6-Trimethyl-1-cyclohexenyl)-2-buten-1-one
Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Chemical Composition
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade
Showing 1-3 of 3