Skip to Content
Merck
Disulfoton-sulfone PESTANAL®, analytical standard

Disulfoton-sulfone

Disulfoton-sulfone PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C8H19O4PS3
CAS Number:
Molecular Weight:
306.40

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique