Skip to Content
Merck
D-Isoascorbic acid FG

D-(−)-Isoascorbic acid

D-Isoascorbic acid FG
Synonym(s):
D-erythro-Hex-2-enoic acid γ-lactone, D-Araboascorbic acid, Erythorbic acid, Glucosaccharonic acid, NSC 8117, D-(−)-Isoascorbic acid
Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing