Skip to Content
Merck
vibrant-m

Diprenorphine

vibrant-m
Synonym(s):
(5α,7α)-17-(Cyclopropylmethyl)-4,5-epoxy-18,19-dihydro-3-hydroxy-6-methoxy-α,α-dimethyl-6,14-ethenomorphinan-7-methanol
Empirical Formula (Hill Notation):
C26H35NO4
CAS Number:
Molecular Weight:
425.56
EC Number:

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing