Deferasirox impurity F
Synonym(s):
4-Hydrazinobenzoic acid
Linear Formula:
H2NNHC6H4CO2H
CAS Number:
Molecular Weight:
152.15
Products
Formula Weight
Manufacturer
Melting Point (°C)
Markush Class
Markush Group
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing