Skip to Content
Merck
D-(+)-Cellobiose analytical standard

D-(+)-Cellobiose

D-(+)-Cellobiose analytical standard
Synonym(s):
β-D-Glc-(1→4)-D-Glc, 4-O-β-D-Glucopyranosyl-D-glucose
Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Analyte Suitability
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Technique