Skip to Content
Merck
Cyclopentanone analytical standard

Cyclopentanone

Cyclopentanone analytical standard
Synonym(s):
Cyclopentanone
Linear Formula:
C5H8(=O)
CAS Number:
Molecular Weight:
84.12
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing