Skip to Content
Merck
Cinnamyl alcohol ≥98%, FG

Cinnamyl alcohol

Cinnamyl alcohol ≥98%, FG
Synonym(s):
3-Phenyl-2-propen-1-ol
Linear Formula:
C6H5CH=CHCH2OH
CAS Number:
Molecular Weight:
134.18
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing