![Carbachol European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/186/980/1435bbde-ba48-47b6-9724-7d1190b7415d/640/1435bbde-ba48-47b6-9724-7d1190b7415d.png)
Carbachol
![Carbachol European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/186/980/1435bbde-ba48-47b6-9724-7d1190b7415d/640/1435bbde-ba48-47b6-9724-7d1190b7415d.png)
Synonym(s):
Carbamoylcholine chloride, (2-Hydroxyethyl)trimethylammonium chloride carbamate, Carbachol, Carbamylcholine chloride
Linear Formula:
NH2COOCH2CH2N(Cl)(CH3)3
CAS Number:
Molecular Weight:
182.65
Products
Shipping
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Technique
Showing 1-3 of 3