Skip to Content
Merck
Butyl propionate ≥98%, FG

Butyl propionate

Butyl propionate ≥98%, FG
Linear Formula:
C2H5CO2(CH2)3CH3
CAS Number:
Molecular Weight:
130.18
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing