Skip to Content
Merck
Boc-Met-OH 99%

Boc-Met-OH

Boc-Met-OH 99%
Synonym(s):
N-(tert-Butoxycarbonyl)-L-methionine, Boc-L-methionine, Boc-Met-OH
Linear Formula:
CH3SCH2CH2CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
249.33

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing