Skip to Content
Merck
Benzyl Nicotinate Pharmaceutical Secondary Standard; Certified Reference Material

Benzyl nicotinate

Benzyl Nicotinate Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Nicotinic acid benzyl ester, Benzyl nicotinate
Empirical Formula (Hill Notation):
C13H11NO2
CAS Number:
Molecular Weight:
213.23
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2