Skip to Content
Merck
Ascentis® C8 HPLC Column 5 μm particle size, L × I.D. 15 cm × 4.6 mm

Ascentis® C8 (5 µm) HPLC Columns

Ascentis® C8 HPLC Column 5 μm particle size, L × I.D. 15 cm × 4.6 mm

Products

Shipping
Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Application
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique