Skip to Content
Merck
Anti-XYLT1 antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-XYLT1 antibody produced in rabbit

Anti-XYLT1 antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-Peptide O-xylosyltransferase 1 antibody produced in rabbit, Anti-XT-I antibody produced in rabbit, Anti-XylT-I antibody produced in rabbit, Anti-Xylosyltransferase 1 antibody produced in rabbit, Anti-Xylosyltransferase I antibody produced in rabbit

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique