
Anti-γ-Tubulin Antibody

Synonym(s):
Anti-γ-Tubulin, γ-1-tubulin, γ-tubulin complex component 1, GCP-1, TUBG1, Tubulin γ-1 chain
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Package Size
Post Translational Modification
Species Reactivity
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing