Anti-PRRC1 antibody produced in rabbit
Synonym(s):
Anti-Flj32875, Anti-Proline-rich coiled-coil 1
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1