Anti-MASP1 antibody produced in rabbit
Synonym(s):
Anti-Complement-activating component of Ra-reactive factor precursor antibody produced in rabbit, Anti-MASP-1 antibody produced in rabbit, Anti-Mannan-binding lectin serine protease 1 antibody produced in rabbit, Anti-Mannose-binding protein-associated serine protease antibody produced in rabbit, Anti-Ra-reactive factor serine protease p100 antibody produced in rabbit, Anti-RaRF antibody produced in rabbit, Anti-Serine protease 5 antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing