
Androstenedione

Synonym(s):
4-Androstene-3,17-dione, Androstenedione
Empirical Formula (Hill Notation):
C19H26O2
CAS Number:
Molecular Weight:
286.41
Products
Shipping
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1